Merck
  • Potent neutralization of SARS-CoV-2 by human antibody heavy-chain variable domains isolated from a large library with a new stable scaffold.
Potent neutralization of SARS-CoV-2 by human antibody heavy-chain variable domains isolated from a large library with a new stable scaffold.
mAbs (2020-06-17)
Zehua Sun, Chuan Chen, Wei Li, David R Martinez, Aleksandra Drelich, Du-San Baek, Xianglei Liu, John W Mellors, Chien-Te Tseng, Ralph S Baric, Dimiter S Dimitrov
ABSTRACT
Effective therapies are urgently needed for COVID-19. Here we describe the identification of a new stable human immunoglobulin G1 heavy-chain variable (VH) domain scaffold that was used for the construction of a large library, lCAT6, of engineered human VHs. This library was panned against the receptor-binding domain (RBD) of the SARS-CoV-2 spike (S) glycoprotein. Two VH domains (VH ab6 and VH m397) were selected and fused to Fc for increased half-life in circulation. The VH-Fc ab6 and m397 specifically neutralized SARS-CoV-2 with high potencies (50% neutralization at 0.35 µg/ml and 1.5 µg/ml, respectively) as measured by two independent replication-competent virus neutralization assays. Ab6 and m397 competed with ACE2 for binding to RBD, suggesting a competitive mechanism of virus neutralization. These VH domains may have potential applications for prophylaxis and therapy of COVID-19 alone or in combination, as well as for diagnosis and as tools for research.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Human IgG (γ-chain specific)−R-Phycoerythrin antibody produced in goat, affinity isolated antibody, buffered aqueous solution