Merck
  • Concentrations of bile acid precursors in cerebrospinal fluid of Alzheimer's disease patients.
Concentrations of bile acid precursors in cerebrospinal fluid of Alzheimer's disease patients.
Free radical biology & medicine (2018-12-24)
William J Griffiths, Jonas Abdel-Khalik, Eylan Yutuc, Gustavo Roman, Margaret Warner, Jan-Åke Gustafsson, Yuqin Wang
ABSTRACT
Using liquid chromatography - mass spectrometry in combination with derivatisation chemistry we profiled the oxysterol and cholestenoic acid content of cerebrospinal fluid from patients with Alzheimer's disease (n = 21), vascular dementia (n = 11), other neurodegenerative diseases (n = 15, Lewy bodies dementia, n = 3, Frontotemporal dementia, n = 11) and controls (n = 15). Thirty different sterols were quantified and the bile acid precursor 7α,25-dihydroxy-3-oxocholest-4-en-26-oic acid found to be reduced in abundance in cerebrospinal fluid of Alzheimer's disease patient-group. This was the only sterol found to be changed amongst the different groups.
Product Description
Avanti
3β,7α,25-trihydroxy-5-cholestenoic acid, Avanti Research - A Croda Brand 700132P, powder