Merck
  • Part I. Polyphenols composition and antioxidant potential during 'Blaufränkisch' grape maceration and red wine maturation, and the effects of trans-resveratrol addition.
Part I. Polyphenols composition and antioxidant potential during 'Blaufränkisch' grape maceration and red wine maturation, and the effects of trans-resveratrol addition.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2020-01-14)
Nataša Poklar Ulrih, Rok Opara, Mihaela Skrt, Tatjana Košmerl, Mojmir Wondra, Veronika Abram
ABSTRACT
The effects of extended maceration (13 days) were investigated for extraction of trans-resveratrol and other phenolics from grapes of cultivar 'Blaufränkisch', and then during the subsequent maturation of the wine (250 days). Total phenolics and three subgroups of phenolics were followed. The concentrations of total phenolics, total flavonoids, and total anthocyanins, but not of total nonflavonoids, increased with extended maceration, and then decreased after 250 days of maturation. Trans- and cis-resveratrol concentrations increased following extended maceration and maturation (6.5, 2.9 mg/L, respectively). The maximum polydatin concentration was reached after only 6 days of maceration (10.9 mg/L). The antioxidant potential of the must increased following extended maceration (12.3 mmol DPPH2/L), and then remained unchanged for the red wine after maturation. Addition of trans-resveratrol to the red wine and into model solutions showed increased solubility and stability of trans-resveratrol in the red wines over the model solutions. Minor increases in antioxidant potential and better stability of malvidin-3-glucoside were also seen.
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)