- Like-charge interactions between colloidal particles are asymmetric with respect to sign.
Like-charge interactions between colloidal particles are asymmetric with respect to sign.
Soft matter (2009-05-07)
Esther W Gomez, Nathan G Clack, Hung-Jen Wu, Jay T Groves
PMID25221611
ABSTRACT
Two-dimensional dispersions of colloidal particles with a range of surface chemistries and electrostatic potentials are characterized under a series of solution ionic strengths. A combination of optical imaging techniques are employed to monitor both the colloid structure and the electrostatic surface potential of individual particles in situ. We find that like-charge multiparticle interactions can be tuned from exclusively repulsive to long-range attractive by changing the particle surface composition. This behavior is strongly asymmetric with respect to the sign of the surface potential. Collective long-range attractive interactions are only observed among negatively charged particles.