Merck
  • Characterization of phosphatidylcholine/polyethylene glycol-lipid aggregates and their use as coatings and carriers in capillary electrophoresis.
Characterization of phosphatidylcholine/polyethylene glycol-lipid aggregates and their use as coatings and carriers in capillary electrophoresis.
Electrophoresis (2008-01-24)
Maria V Lindén, Kristoffer Meinander, Anne Helle, Gebrenegus Yohannes, Marja-Liisa Riekkola, Sarah J Butcher, Tapani Viitala, Susanne K Wiedmer
ABSTRACT
PEG-stabilized lipid aggregates are a promising new class of model membranes in biotechnical and pharmaceutical applications. CE techniques, field-flow fractionation, light scattering, quartz crystal microbalance (QCM), and microscopic techniques were used to study aggregates composed of 1-palmitoyl-2-oleyl-sn-glycero-phosphatidylcholine (POPC) and PEG-lipid conjugates. The PEG-lipids, with PEG molar masses of 1000, 2000, and 3000, were 1,2-diacyl-sn-glycero-3-phosphoethanolamine-N-[methoxy-(PEG)] derivatives with either dimyristoyl (DM, 14:0) or distearoyl (DS, 18:0) acyl groups. The 80/20 mol% POPC/PEG-lipid dispersions in HEPES at pH 7.4 were extruded through 100 nm size membranes. Asymmetrical flow field-flow fractionation (AsFlFFF), photon correlation spectroscopy (PCS), and dynamic light scattering (DLS) were used to determine the sizes of POPC and the PEGylated aggregates. All methods demonstrated that the DSPEG-lipid sterically stabilized aggregates were smaller in size than pure POPC vesicles. The zeta potentials of the aggregates were measured and showed an increase from -19 mV for pure POPC to -4 mV for the POPC/DSPEG3000 aggregates. Atomic force microscopy (AFM), electron cryo-microscopy (EM), and multifrequency QCM studies were made to achieve information about the PEGylated coatings on silica. Lipid aggregates with different POPC/DSPEG3000-lipid ratios were applied as capillary coating material, and the 80/20 mol% composition was found to give the most suppressed and stable EOFs. Mixtures of low-molar-mass drugs and FITC-labeled amino acids were separated with the PEGylated aggregates as carriers (EKC) or as coating material (CEC). Detection was made by UV and LIF.
Product Description
Avanti
14:0 PEG1000 PE, Avanti Research - A Croda Brand 880710P, powder