Merck
  • Early development of calcific aortic valve disease and left ventricular hypertrophy in a mouse model of combined dyslipidemia and type 2 diabetes mellitus.
Early development of calcific aortic valve disease and left ventricular hypertrophy in a mouse model of combined dyslipidemia and type 2 diabetes mellitus.
Arteriosclerosis, thrombosis, and vascular biology (2014-09-19)
Khai Le Quang, Rihab Bouchareb, Dominic Lachance, Marc-André Laplante, Diala El Husseini, Marie-Chloé Boulanger, Dominique Fournier, Xiang Ping Fang, Rita Kohen Avramoglu, Philippe Pibarot, Yves Deshaies, Gary Sweeney, Patrick Mathieu, André Marette
ABSTRACT
This study aimed to determine the potential impact of type 2 diabetes mellitus on left ventricular dysfunction and the development of calcified aortic valve disease using a dyslipidemic mouse model prone to developing type 2 diabetes mellitus. When compared with nondiabetic LDLr(-/-)/ApoB(100/100), diabetic LDLr(-/-)/ApoB(100/100)/IGF-II mice exhibited similar dyslipidemia and obesity but developed type 2 diabetes mellitus when fed a high-fat/sucrose/cholesterol diet for 6 months. LDLr(-/-)/ApoB(100/100)/IGF-II mice showed left ventricular hypertrophy versus C57BL6 but not LDLr(-/-)/ApoB(100/100) mice. Transthoracic echocardiography revealed significant reductions in both left ventricular systolic fractional shortening and diastolic function in high-fat/sucrose/cholesterol fed LDLr(-/-)/ApoB(100/100)/IGF-II mice when compared with LDLr(-/-)/ApoB(100/100). Importantly, we found that peak aortic jet velocity was significantly increased in LDLr(-/-)/ApoB(100/100)/IGF-II mice versus LDLr(-/-)/ApoB(100/100) animals on the high-fat/sucrose/cholesterol diet. Microtomography scans and Alizarin red staining indicated calcification in the aortic valves, whereas electron microscopy and energy dispersive x-ray spectroscopy further revealed mineralization of the aortic leaflets and the presence of inflammatory infiltrates in diabetic mice. Studies showed upregulation of hypertrophic genes (anp, bnp, b-mhc) in myocardial tissues and of osteogenic genes (spp1, bglap, runx2) in aortic tissues of diabetic mice. We have established the diabetes mellitus -prone LDLr(-/-)/ApoB(100/100)/IGF-II mouse as a new model of calcified aortic valve disease. Our results are consistent with the growing body of clinical evidence that the dysmetabolic state of type 2 diabetes mellitus contributes to early mineralization of the aortic valve and calcified aortic valve disease pathogenesis.
Sigma-Aldrich
Anti-Osteopontin Antibody, serum, from rabbit