Merck
  • GRK2 suppresses lymphomagenesis by inhibiting the MALT1 proto-oncoprotein.
GRK2 suppresses lymphomagenesis by inhibiting the MALT1 proto-oncoprotein.
The Journal of clinical investigation (2020-01-22)
Jing Cheng, Linda R Klei, Nathaniel E Hubel, Ming Zhang, Rebekka Schairer, Lisa M Maurer, Hanna B Klei, Heejae Kang, Vincent J Concel, Phillip C Delekta, Eric V Dang, Michelle A Mintz, Mathijs Baens, Jason G Cyster, Narayanan Parameswaran, Margot Thome, Peter C Lucas, Linda M McAllister-Lucas
ABSTRACT
Antigen receptor-dependent (AgR-dependent) stimulation of the NF-κB transcription factor in lymphocytes is a required event during adaptive immune response, but dysregulated activation of this signaling pathway can lead to lymphoma. AgR stimulation promotes assembly of the CARMA1-BCL10-MALT1 complex, wherein MALT1 acts as (a) a scaffold to recruit components of the canonical NF-κB machinery and (b) a protease to cleave and inactivate specific substrates, including negative regulators of NF-κB. In multiple lymphoma subtypes, malignant B cells hijack AgR signaling pathways to promote their own growth and survival, and inhibiting MALT1 reduces the viability and growth of these tumors. As such, MALT1 has emerged as a potential pharmaceutical target. Here, we identified G protein-coupled receptor kinase 2 (GRK2) as a new MALT1-interacting protein. We demonstrated that GRK2 binds the death domain of MALT1 and inhibits MALT1 scaffolding and proteolytic activities. We found that lower GRK2 levels in activated B cell-type diffuse large B cell lymphoma (ABC-DLBCL) are associated with reduced survival, and that GRK2 knockdown enhances ABC-DLBCL tumor growth in vitro and in vivo. Together, our findings suggest that GRK2 can function as a tumor suppressor by inhibiting MALT1 and provide a roadmap for developing new strategies to inhibit MALT1-dependent lymphomagenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting human GRK2
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)