Merck
  • Different Complicated Brain Pathologies in Monozygotic Twins With Gerstmann-Sträussler-Scheinker Disease.
Different Complicated Brain Pathologies in Monozygotic Twins With Gerstmann-Sträussler-Scheinker Disease.
Journal of neuropathology and experimental neurology (2017-09-20)
Hiroyuki Honda, Kensuke Sasaki, Hiroshi Takashima, Daisuke Mori, Sachiko Koyama, Satoshi O Suzuki, Toru Iwaki
ABSTRACT
Gerstmann-Sträussler-Scheinker disease (GSS) is an autosomal, dominantly inherited prion disease. In this study, we present different complicated brain pathologies determined postmortem of monozygotic GSS twin sisters. Case 1 showed cerebellar ataxia at the age of 58 years, and died at 66 years. Case 2 became symptomatic at the age of 75 years, and died at 79 years. There was a 17-year difference in the age of onset between the twins. Postmortem examination revealed numerous prion protein (PrP) plaques in the brains of both cases. The spongiform change and brain atrophy in case 1 were more severe compared with those in case 2. Western-blot analysis identified proteinase-resistant PrP (PrPres) at the molecular weight of 21-30 kDa and 8 kDa in the twins. Gel filtration revealed that PrPres was mainly composed of PrP oligomer. PrPres signal patterns were similar between the twins. Additionally, case 1 showed α-synucleinopathy and case 2 showed Alzheimer disease pathology. These different proteinopathies were involved in the amyloid plaque formations of both cases. The degree of GSS pathology was mainly related to disease duration. The amyloid plaque formations could be decorated by concomitant neuropathological changes such as α-synucleinopathy and tauopathy.
Sigma-Aldrich
Anti-Tau (4-repeat isoform RD4) Antibody, clone 1E1/A6, culture supernatant, clone 1E1/A6, Upstate®
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-Tau (3-repeat isoform RD3) Antibody, clone 8E6/C11, culture supernatant, clone 8E6/C11, Upstate®