Merck
  • Survival response to B-cell receptor ligation is restricted to progressive chronic lymphocytic leukemia cells irrespective of Zap70 expression.
Survival response to B-cell receptor ligation is restricted to progressive chronic lymphocytic leukemia cells irrespective of Zap70 expression.
Cancer research (2006-07-20)
Pierre-Antoine Deglesne, Nathalie Chevallier, Rémi Letestu, Fanny Baran-Marszak, Taoufik Beitar, Célia Salanoubat, Laurence Sanhes, Joelle Nataf, Claudine Roger, Nadine Varin-Blank, Florence Ajchenbaum-Cymbalista
ABSTRACT
Despite very similar gene expression profiles, the clinical course of B-cell chronic lymphocytic leukemia (B-CLL) is heterogeneous. Immunoglobulin VH (IgVH) mutational status and expression of B-cell receptor (BCR) signaling mediators have been associated with disease progression. However, the consequences of BCR engagement on cell survival and evolution of the disease remain unclear. We show here that B-CLL cell survival is dependent on the threshold of BCR stimulation induced by immobilized antibody, in contrast to soluble anti-mu F(ab)'2 antibody, which leads to apoptosis. Measurement of metabolic activity and apoptotic response discriminated two subgroups. "Nonresponders" showed low metabolic activity and unmodified apoptotic response upon BCR stimulation. In contrast, "responders" exhibited increased metabolic activity and inhibition of spontaneous apoptosis. This survival advantage was associated to a BCR-dependent activation profile leading to induction of cyclin D2/cyclin-dependent kinase 4 (cdk4) expression and G1 cell cycle progression. The ability to respond to BCR ligation correlated with an unfavorable clinical course and allowed to define an additional group of patients among IgVH-mutated cases exhibiting a risk of progression. Remarkably, we show that Zap70 expression was neither mandatory nor sufficient to generate downstream survival signals and cyclin D2/cdk4 up-regulation. In conclusion, BCR engagement has a significant effect on B-CLL cell survival, activation, and G1 progression. Furthermore, our results provide new insights in the physiopathology of progressive IgVH-mutated cases.
Sigma-Aldrich
Anti-ZAP-70 Antibody, clone 2F3.2, clone 2F3.2, Upstate®, from mouse