Merck
  • Mifepristone-inducible caspase-1 expression in mouse embryonic stem cells eliminates tumor formation but spares differentiated cells in vitro and in vivo.
Mifepristone-inducible caspase-1 expression in mouse embryonic stem cells eliminates tumor formation but spares differentiated cells in vitro and in vivo.
Stem cells (Dayton, Ohio) (2011-12-02)
Yi Wang, Dehua Yang, Lin Song, Ting Li, Juan Yang, Xiaojie Zhang, Weidong Le
ABSTRACT
Embryonic stem cell (ESC)-based therapy is a promising treatment for neurodegenerative diseases. But there is always a risk of tumor formation that is due to contamination of undifferentiated ESCs. To reduce the risk and improve ESC-based therapy, we have established a novel strategy by which we can selectively eliminate tumor cells derived from undifferentiated ESCs but spare differentiated cells. In this study, we generated a caspase-1-ESC line transfected with a mifepristone-regulated caspase-1 expression system. Mifepristone induced caspase-1 overexpression both in differentiated and undifferentiated caspase-1-ESCs. All the undifferentiated caspase-1-ESCs were induced to death after mifepristone treatment. Tumors derived from undifferentiated caspase-1-ESCs were eliminated following 3 weeks of mifepristone treatment in vivo. However, differentiated caspase-1-ESCs survived well under the condition of mifepristone-induced caspase-1 overexpression. To examine in vivo the impact of mifepristone-induced caspase-1 activation on grafted cells, we transplanted wild-type ESCs or caspase-1-ESCs into nude mice brains. After 8 weeks of mifepristone treatment, we could not detect any tumor cells in the caspase-1-ESC grafts in the brains of mice. However, we found that donor dopamine neurons survived in the recipient brains. These data demonstrate that mifepristone-induced caspase-1 overexpression in ESCs can eliminate the potential tumor formation meanwhile spares the differentiated cells in the host brains. These results suggest that this novel ESC-based therapy can be used in Parkinson's disease and other related disorders without the risk of tumor formation.
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, clone rat-401, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit