Merck
  • Plasma and Liver Protein Binding of N-Acetylgalactosamine-Conjugated Small Interfering RNA.
Plasma and Liver Protein Binding of N-Acetylgalactosamine-Conjugated Small Interfering RNA.
Drug metabolism and disposition: the biological fate of chemicals (2019-05-18)
Sara C Humphreys, Mai B Thayer, Julie M Lade, Bin Wu, Kelvin Sham, Babak Basiri, Yue Hao, Xin Huang, Richard Smith, Brooke M Rock
ABSTRACT
Understanding small interfering RNA (siRNA) fraction unbound (f u) in relevant physiologic compartments is critical for establishing pharmacokinetic-pharmacodynamic relationships for this emerging modality. In our attempts to isolate the equilibrium free fraction of N-acetylgalactosamine-conjugated siRNA using classic small-molecule in vitro techniques, we found that the hydrodynamic radius was critical in determining the size exclusion limit requirements for f u isolation, largely validating the siRNA "rigid rod" hypothesis. With this knowledge, we developed an orthogonally validated 50 kDa molecular-mass cutoff ultrafiltration assay to quantify f u in biologic matrices including human, nonhuman primate, rat, and mouse plasma, and human liver homogenate. To enhance understanding of the siRNA-plasma interaction landscape, we examined the effects of various common oligonucleotide therapeutic modifications to the ribose and helix backbone on siRNA f u in plasma (f u,plasma) and found that chemical modifications can alter plasma protein binding by at least 20%. Finally, to gain insight into which specific plasma proteins bind to siRNA, we developed a qualitative screen to identify binding "hits" across a panel of select purified human plasma proteins.
Sigma-Aldrich
Human IgG, Fc Fragment
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
CHAPS hydrate, ≥98% (HPLC)