Merck
  • Synthesis and biological evaluation of sulfonylhydrazone-substituted imidazo[1,2-a]pyridines as novel PI3 kinase p110alpha inhibitors.
Synthesis and biological evaluation of sulfonylhydrazone-substituted imidazo[1,2-a]pyridines as novel PI3 kinase p110alpha inhibitors.
Bioorganic & medicinal chemistry (2007-07-03)
Masahiko Hayakawa, Ken-Ichi Kawaguchi, Hiroyuki Kaizawa, Tomonobu Koizumi, Takahide Ohishi, Mayumi Yamano, Minoru Okada, Mitsuaki Ohta, Shin-Ichi Tsukamoto, Florence I Raynaud, Peter Parker, Paul Workman, Michael D Waterfield
ABSTRACT
We have previously reported the imidazo[1,2-a]pyridine derivative 4 as a novel p110alpha inhibitor; however, although 4 is a potent inhibitor of p110alpha enzymatic activity and tumor cell proliferation in vitro, it is unstable in solution and ineffective in vivo. To increase stability the pyrazole of 4 was replaced with a hydrazone and a moderately potent p110alpha inhibitor 7a was obtained. Subsequent optimization of 7a afforded exceptionally potent p110alpha inhibitors, including 8c and 8h, with IC(50) values of 0.30 nM and 0.26 nM, respectively; to the best of our knowledge, these compounds are the most potent PI3K p110alpha inhibitors reported to date. Compound 8c was also stable in solution and exhibited significant anti-tumor effectiveness in vivo.
Sigma-Aldrich
PI 3-Kα Inhibitor VIII, The PI 3-Kα Inhibitor VIII controls the biological activity of PI 3-Kα. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.