Merck
  • Synthesis of 3-(Cyclohexylamino)-2-arylimidazo[1,2-a]pyridine-8- carboxylic Acids Via an Efficient Three-component Condensation Reaction between Cyclohexylisocyanide and 2-Aminopyridine-3-carboxylic Acid in the Presence of Aromatic Aldehyde.
Synthesis of 3-(Cyclohexylamino)-2-arylimidazo[1,2-a]pyridine-8- carboxylic Acids Via an Efficient Three-component Condensation Reaction between Cyclohexylisocyanide and 2-Aminopyridine-3-carboxylic Acid in the Presence of Aromatic Aldehyde.
Combinatorial chemistry & high throughput screening (2018-04-18)
Ghasem Marandi
ABSTRACT
The reaction of cyclohexylisocyanide and 2-aminopyridine-3- carboxylic acid in the presence of benzaldehyde derivatives in ethanol led to 3-(cyclohexylamino)-2- arylimidazo[1,2-a]pyridine-8-carboxylic acids in high yields. In a three component condensation reaction, isocyanide reacts with 2-aminopyridine-3-carboxylic acid and aromatic aldehydes without any prior activation. The synthesized products have stable structures which have been characterized by IR, 1H, 13C and Mass spectroscopy as well as CHN-O analysis. In continuation of our attempts to develop simple one-pot routes for the synthesis of 3- (cyclohexylamino)-2-arylimidazo[1,2-a]pyridine-8-carboxylic acids, aromatic aldehydes with divers substituted show a high performance. In conclusion, this study introduces the art of combinatorial chemistry using a simple one-pot procedure for the synthesis of new materials which are interesting compounds in medicinal and biological sciences.