Merck
  • Synthesis, biological properties and anti-HIV-1 activity of new pyrimidine P1,P2-dinucleotides.
Synthesis, biological properties and anti-HIV-1 activity of new pyrimidine P1,P2-dinucleotides.
Nucleosides, nucleotides & nucleic acids (2010-06-15)
A Miazga, P Ziemkowski, M A Siwecka, A Lipniacki, A Piasek, T Kulikowski
ABSTRACT
New homo- and hetero-P(1),P(2)-dinucleotides were prepared with the use of multistep procedures starting from the monophosphates of 3'-fluoro-2-thiothymidine, 3'-fluoro-4-thiothymidine, AZT and 1-[(2-hydroxyethoxy)-methyl-5-propyl-6-phenylselenenyl]uracil. Anti-HIV properties of the synthesized P(1),P(2)-dinucleotides were evaluated against laboratory syncytia inducing strain HIV-1 in CEM-T4 cells. Anti-HIV activities were in the range of 5-45 nM, and therapeutic indexes were higher than 4666-14000. Interactions of the above mentioned compounds with recombinant HIV-1 reverse transcriptase were also investigated. The obtained results point to reverse transcriptase inhibition, with somewhat lower inhibitory activity than that of their parental nucleoside-5'-triphosphates. Compound 6 may be regarded as a potent anti-HIV/AIDS drug.