Merck
  • The mechanisms of citrate on regulating the distribution of carbon flux in the biosynthesis of uridine 5'-monophosphate by Saccharomyces cerevisiae.
The mechanisms of citrate on regulating the distribution of carbon flux in the biosynthesis of uridine 5'-monophosphate by Saccharomyces cerevisiae.
Applied microbiology and biotechnology (2009-10-15)
Yong Chen, Shuya Li, Jian Xiong, Zhenjiang Li, Jianxin Bai, Lei Zhang, Qi Ye, Pingkai Ouyang, Hanjie Ying
ABSTRACT
A whole cell biocatalytic process for uridine 5'-monophosphate (UMP) production from orotic acid by Saccharomyces cerevisiae was developed. The concentration of UMP was increased by 23% when 1 g l(-1) sodium citrate was fed into the broth. Effects of citrate addition on UMP production were investigated. Glucose-6-phosphate pool was elevated by onefold, while FBP and pyruvate were decreased by 42% and 40%, respectively. Organic acid pools such as acetate and succinate were averagely decreased by 30% and 49%. The results demonstrated that manipulation of citrate levels could be used as a novel tool to regulate the metabolic fluxes distribution among glycolysis, pentose phosphate pathway, and TCA cycle.
Sigma-Aldrich
Orotic acid, ≥98% (titration), anhydrous
Sigma-Aldrich
Orotic acid potassium salt, ≥98%