Merck
SnapShot: RNA Structure Probing Technologies.
Cell (2018-10-06)
Paul D Carlson, Molly E Evans, Angela M Yu, Eric J Strobel, Julius B Lucks
ABSTRACT
Chemical probing coupled to high-throughput sequencing offers a flexible approach to uncover many aspects of RNA structure relevant to its cellular function. With a wide variety of chemical probes available that each report on different features of RNA molecules, a broad toolkit exists for investigating in vivo and in vitro RNA structure and interactions with other molecules.
Sigma-Aldrich
1-Methyl-7-nitroisatoic anhydride
Sigma-Aldrich
2-(Azidomethyl)nicotinic acid imidazolide, ≥95%
Sigma-Aldrich
2-Methylnicotinic acid imidazolide