Merck
  • High resolution of multiple forms of rabbit reticulocyte hexokinase type I by hydrophobic interaction chromatography.
High resolution of multiple forms of rabbit reticulocyte hexokinase type I by hydrophobic interaction chromatography.
Journal of chromatography. A (1994-07-29)
V Stocchi, P Cardoni, P Ceccaroli, G Piccoli, L Cucchiarini, R De Bellis, M Dachà
ABSTRACT
Hydrophobic interaction chromatography (HIC) has been employed extensively in the separation of proteins by elution using a descending salt gradient, with and without the use of detergents or denaturing agents. In this study, a new hydrophobic interaction chromatographic support, Toyopearl Phenyl 650 S, was investigated in order to examine the distribution of multiple forms of rabbit reticulocyte hexokinase type I. These distinct forms of the enzyme, designated hexokinase Ia, Ia* and Ib, show similar kinetic and physical properties, similar molecular masses (ca. 100,000) and a different intracellular distribution. The results obtained using Toyopearl Phenyl 650 S of 20-50-microns particle diameter show that this HIC support allows very high resolution, comparable to that obtainable with HIC-HPLC columns but with the advantage of charging a higher amount of starting material even with a high protein concentration. These characteristics render Toyopearl Phenyl 650 S suitable for analytical and preparative purposes. Further, in the separation of multiple forms of rabbit reticulocyte hexokinase, the HIC method was shown to be superior to RP-HPLC, making possible the efficient separation of proteins with high molecular mass and their recovery in active forms. The Toyopearl Phenyl 650 S column was also shown to be more efficient than the ion-exchange chromatographic media previously used, allowing a quicker analysis of the multiple forms of rabbit reticulocyte hexokinase under different biological conditions.
Product Description
TOYOPEARL® Butyl-650M Bulk Media, matrix polymer (65 μm), bottle of 100 mL, 65 μm
TOYOPEARL® Phenyl-650M Bulk Media, matrix polymer (65 μm), bottle of 100 mL, 65 μm