Merck
  • Practical and Broadly Applicable Catalytic Enantioselective Additions of Allyl-B(pin) Compounds to Ketones and α-Ketoesters.
Practical and Broadly Applicable Catalytic Enantioselective Additions of Allyl-B(pin) Compounds to Ketones and α-Ketoesters.
Angewandte Chemie (International ed. in English) (2016-06-09)
Daniel W Robbins, KyungA Lee, Daniel L Silverio, Alexey Volkov, Sebastian Torker, Amir H Hoveyda
ABSTRACT
A set of broadly applicable methods for efficient catalytic additions of easy-to-handle allyl-B(pin) (pin=pinacolato) compounds to ketones and acyclic α-ketoesters was developed. Accordingly, a large array of tertiary alcohols can be obtained in 60 to >98 % yield and up to 99:1 enantiomeric ratio. At the heart of this development is rational alteration of the structures of the small-molecule aminophenol-based catalysts. Notably, with ketones, increasing the size of a catalyst moiety (tBu to SiPh3 ) results in much higher enantioselectivity. With α-ketoesters, on the other hand, not only does the opposite hold true, since Me substitution leads to substantially higher enantioselectivity, but the sense of the selectivity is reversed as well.