- [Determination of dimethylbenzoic acid isomers in urine by gas chromatography].
[Determination of dimethylbenzoic acid isomers in urine by gas chromatography].
Trimethylobenzene (TMB) is a main ingredient of many organic solvents used in industry. In Farbasol (Polish trade name of the solvent) TMB occurs as a mixture of three isomers: pseudocumene (1, 2, 4-TMB) 30%; mesitylene (1, 3, 5-TMB) 15%; hemimellitene (1,2,3-TMB) 5%. As it is known in human organism, TMB is metabolized mainly to dimethylbenzoic (DMBA) and dimethylhippuric (DMHA) acids, and some authors suggest, that the acids excreted in urine can be biological indicators of exposure to TMB. This study was aimed at developing the method of determination of DMBA isomers in urine. Biological material was hydrolyzed with sodium hydroxide and next extracted with diethyl ether. DMBA concentration in urine was determined by gas chromatography using a variant of quantitative analysis with internal standard (5-methyl-2-isopropylphenol, thymol). Analytical parameters of the developed method of determination of DMBA isomers in urine such as linearity, precision, reproducibility, stability (192 days, when urine samples stored at-18 degrees C), detectability limit (400 micrograms/dm3) have been fully compatible with the requirements of biological monitoring. In order to confirm the presence of DMBA isomers in urine, four volunteers were exposed (8 hours) to Farbasol in toxicological chamber. The TMB concentration in the air, determined by means of gas chromatograph (HP 5890), amounted to 100 mg/m3 (MAC value in Poland). In urine samples collected 2,3-; 2,4-; 2,5-; 2,6-; 3,4-; 3,5-dimethylbenzoic acids were identified by means of GC/MSD.(ABSTRACT TRUNCATED AT 250 WORDS)