Merck
  • Interactions of non-polar and "Click-able" nucleotides in the confines of a DNA polymerase active site.
Interactions of non-polar and "Click-able" nucleotides in the confines of a DNA polymerase active site.
Chemical communications (Cambridge, England) (2012-07-07)
Samra Obeid, Holger Busskamp, Wolfgang Welte, Kay Diederichs, Andreas Marx
ABSTRACT
Modified nucleotides play a paramount role in many cutting-edge biomolecular techniques. The present structural study highlights the plasticity and flexibility of the active site of a DNA polymerase while incorporating non-polar "Click-able" nucleotide analogs and emphasizes new insights into rational design guidelines for modified nucleotides.
Sigma-Aldrich
BTTAA, ≥95%