Merck
  • The autophagy inhibitor spautin-1, either alone or combined with doxorubicin, decreases cell survival and colony formation in canine appendicular osteosarcoma cells.
The autophagy inhibitor spautin-1, either alone or combined with doxorubicin, decreases cell survival and colony formation in canine appendicular osteosarcoma cells.
PloS one (2018-10-30)
Courtney R Schott, Latasha Ludwig, Anthony J Mutsaers, Robert A Foster, Geoffrey A Wood
ABSTRACT
Dogs diagnosed with appendicular osteosarcoma typically succumb to metastatic disease within a year of diagnosis. The current standard of care for curative intent, amputation followed by adjuvant chemotherapy, increases survival time but chemoresistance is a major contributor to mortality. Unfortunately, the mechanisms driving the progression of metastatic disease and the development of chemoresistance are unknown. One theory is that autophagy may contribute to chemoresistance by providing neoplastic cells with a mechanism to survive chemotherapy treatment. Our objective was to evaluate the effect of combining an autophagy inhibitor with a standard chemotherapeutic drug on response to chemotherapy in canine appendicular osteosarcoma cells. We hypothesized that combining the autophagy inhibitor spautin-1 with doxorubicin treatment would enhance chemoresponsiveness. Using commercial (D17) and primary cell lines derived from 1° and 2° sites of osteosarcoma, we showed that this combination treatment enhances cell killing and inhibits colony formation. Our findings support the theory that autophagy contributes to chemoresistance in canine appendicular osteosarcoma and indicate that adding an autophagy inhibitor to the standard of care has the potential to improve outcome.
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Gentamicin solution, 10 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Spautin-1, ≥98% (HPLC)