Merck
The Role of m6A/m-RNA Methylation in Stress Response Regulation.
Neuron (2018-07-27)
Mareen Engel, Carola Eggert, Paul M Kaplick, Matthias Eder, Simone Röh, Lisa Tietze, Christian Namendorf, Janine Arloth, Peter Weber, Monika Rex-Haffner, Shay Geula, Mira Jakovcevski, Jacob H Hanna, Dena Leshkowitz, Manfred Uhr, Carsten T Wotjak, Mathias V Schmidt, Jan M Deussing, Elisabeth B Binder, Alon Chen
ABSTRACT
N6-methyladenosine (m6A) and N6,2'-O-dimethyladenosine (m6Am) are abundant mRNA modifications that regulate transcript processing and translation. The role of both, here termed m6A/m, in the stress response in the adult brain in vivo is currently unknown. Here, we provide a detailed analysis of the stress epitranscriptome using m6A/m-seq, global and gene-specific m6A/m measurements. We show that stress exposure and glucocorticoids region and time specifically alter m6A/m and its regulatory network. We demonstrate that deletion of the methyltransferase Mettl3 or the demethylase Fto in adult neurons alters the m6A/m epitranscriptome, increases fear memory, and changes the transcriptome response to fear and synaptic plasticity. Moreover, we report that regulation of m6A/m is impaired in major depressive disorder patients following glucocorticoid stimulation. Our findings indicate that brain m6A/m represents a novel layer of complexity in gene expression regulation after stress and that dysregulation of the m6A/m response may contribute to the pathophysiology of stress-related psychiatric disorders.
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Anti-FTO Antibody, clone 5-2H10, clone 5-2H10, from mouse
Sigma-Aldrich
Corticosterone: HBC complex, free flowing powder