Merck
  • A NUMB-EFA6B-ARF6 recycling route controls apically restricted cell protrusions and mesenchymal motility.
A NUMB-EFA6B-ARF6 recycling route controls apically restricted cell protrusions and mesenchymal motility.
The Journal of cell biology (2018-08-01)
Martina Zobel, Andrea Disanza, Francesca Senic-Matuglia, Michel Franco, Ivan Nicola Colaluca, Stefano Confalonieri, Sara Bisi, Elisa Barbieri, Giusi Caldieri, Sara Sigismund, Salvatore Pece, Philippe Chavrier, Pier Paolo Di Fiore, Giorgio Scita
ABSTRACT
The endocytic protein NUMB has been implicated in the control of various polarized cellular processes, including the acquisition of mesenchymal migratory traits through molecular mechanisms that have only been partially defined. Here, we report that NUMB is a negative regulator of a specialized set of understudied, apically restricted, actin-based protrusions, the circular dorsal ruffles (CDRs), induced by either PDGF or HGF stimulation. Through its PTB domain, NUMB binds directly to an N-terminal NPLF motif of the ARF6 guanine nucleotide exchange factor, EFA6B, and promotes its exchange activity in vitro. In cells, a NUMB-EFA6B-ARF6 axis regulates the recycling of the actin regulatory cargo RAC1 and is critical for the formation of CDRs that mark the acquisition of a mesenchymal mode of motility. Consistently, loss of NUMB promotes HGF-induced cell migration and invasion. Thus, NUMB negatively controls membrane protrusions and the acquisition of mesenchymal migratory traits by modulating EFA6B-ARF6 activity.
Sigma-Aldrich
Anti-PSD4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
MISSION® esiRNA, targeting human NUMB
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® esiRNA, targeting human PSD4