Merck
  • 14-3-3 protein binds to insulin receptor substrate-1, one of the binding sites of which is in the phosphotyrosine binding domain.
14-3-3 protein binds to insulin receptor substrate-1, one of the binding sites of which is in the phosphotyrosine binding domain.
The Journal of biological chemistry (1997-10-06)
T Ogihara, T Isobe, T Ichimura, M Taoka, M Funaki, H Sakoda, Y Onishi, K Inukai, M Anai, Y Fukushima, M Kikuchi, Y Yazaki, Y Oka, T Asano
ABSTRACT
Insulin binding to its receptor induces the phosphorylation of cytosolic substrates, insulin receptor substrate (IRS)-1 and IRS-2, which associate with several Src homology-2 domain-containing proteins. To identify unique IRS-1-binding proteins, we screened a human heart cDNA library with 32P-labeled recombinant IRS-1 and obtained two isoforms (epsilon and zeta) of the 14-3-3 protein family. 14-3-3 protein has been shown to associate with IRS-1 in L6 myotubes, HepG2 hepatoma cells, Chinese hamster ovary cells, and bovine brain tissue. IRS-2, a protein structurally similar to IRS-1, was also shown to form a complex with 14-3-3 protein using a baculovirus expression system. The amount of 14-3-3 protein associated with IRS-1 was not affected by insulin stimulation but was increased significantly by treatment with okadaic acid, a potent serine/threonine phosphatase inhibitor. Peptide inhibition experiments using phosphoserine-containing peptides of IRS-1 revealed that IRS-1 contains three putative binding sites for 14-3-3 protein (Ser-270, Ser-374, and Ser-641). Among these three, the motif around Ser-270 is located in the phosphotyrosine binding domain of IRS-1, which is responsible for the interaction with the insulin receptor. Indeed, a truncated mutant of IRS-1 consisting of only the phosphotyrosine binding domain retained the capacity to bind to 14-3-3 protein in vivo. Finally, the effect of 14-3-3 protein binding on the insulin-induced phosphorylation of IRS-1 was investigated. Phosphoamino acid analysis revealed that IRS-1 coimmunoprecipitated with anti-14-3-3 antibody to be weakly phosphorylated after insulin stimulation, on tyrosine as well as serine residues, compared with IRS-1 immunoprecipitated with anti-IRS-1 antibody. Thus, the association with 14-3-3 protein may play a role in the regulation of insulin sensitivity by interrupting the association between the insulin receptor and IRS-1.
Sigma-Aldrich
IRS1 (600-1245), GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
IRS1 (1-355), GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution