Merck
  • The Effect of Negative Pressure Wound Therapy with and without Instillation on Mature Biofilms In Vitro.
The Effect of Negative Pressure Wound Therapy with and without Instillation on Mature Biofilms In Vitro.
Materials (Basel, Switzerland) (2018-05-19)
Shamaila Tahir, Matthew Malone, Honghua Hu, Anand Deva, Karen Vickery
ABSTRACT
To investigate the effect of negative pressure wound therapy (NPWT) with and without instillation (NPWTi) on in vitro mature biofilm. Mature biofilms of Pseudomonas aeruginosa and Staphylococcus aureus were grown under shear (130 rpm) on polycarbonate coupons in a CDC biofilm reactor for 3 days. Coupons containing biofilms were placed in a sterile petri dish and sealed using NPWT or NPWTi. Coupons were exposed to treatment for 24 h with NPWT alone or with instillation of: Povidone iodine solution (PVP-I) (10% w/v equivalent to 1% w/v available iodine, BETADINE®, Mundipharma, Singapore), surfactant based antimicrobial solution with polyhexamethylene biguanide (SBPHMB) (Prontosan®, B Braun Medical, Melsungen, Germany), Gentamicin 1 µg/mL (GM) (G1264 Sigma-Aldrich Pty Ltd., Castle Hill, Australia) Rifampicin 24 µg/mL (RF) (R3501 Sigma-Aldrich Pty Ltd., Castle Hill, Australia) and NaCl 0.9% (Baxter, Deerfield, IL, USA). Bacterial cell viability and biofilm architecture pre-and post-treatment were assessed using colony forming units (cfu), Live/Dead viability staining, confocal laser scanning microscopy (CLSM) and scanning electron microscopy (SEM). Significant reductions were obtained in S. aureus biofilm thickness (65%) and mass (47%) when treated with NPWTi as compared to NPWT only. NPWTi with instillation of SBPHMB, PVP-I and RF achieved between 2 and 8 log10 reductions against S. aureus biofilm (p < 0.05⁻0.001). Conversely, PVP-I and SBMO achieved a 3.5 log10 reduction against P. aeruginosa (p < 0.05). NPWT alters biofilm architecture by reducing biofilm thickness and mass, but this does not affect bacterial cell viability. NPWT with instillation of certain antimicrobials solutions may provide a further synergistic effect in reducing the number of viable biofilm microorganisms. Our in vitro model may be used for screening the effectiveness of antimicrobials used under instillation prior to animal or human studies.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Gentamicin sulfate salt, powder, BioReagent, suitable for cell culture