Merck
  • Acid Sphingomyelinase Promotes Cellular Internalization of Clostridium perfringens Iota-Toxin.
Acid Sphingomyelinase Promotes Cellular Internalization of Clostridium perfringens Iota-Toxin.
Toxins (2018-05-23)
Masahiro Nagahama, Masaya Takehara, Kazuaki Miyamoto, Kazumi Ishidoh, Keiko Kobayashi
ABSTRACT
Clostridium perfringens iota-toxin is a binary actin-ADP-ribosylating toxin composed of the enzymatic component Ia and receptor binding component Ib. Ib binds to a cell surface receptor, forms Ib oligomer in lipid rafts, and associates with Ia. The Ia-Ib complex then internalizes by endocytosis. Here, we showed that acid sphingomyelinase (ASMase) facilitates the cellular uptake of iota-toxin. Inhibitions of ASMase and lysosomal exocytosis by respective blockers depressed cell rounding induced by iota-toxin. The cytotoxicity of the toxin increased in the presence of Ca2+ in extracellular fluids. Ib entered target cells in the presence but not the absence of Ca2+. Ib induced the extracellular release of ASMase in the presence of Ca2+. ASMase siRNA prevented the cell rounding induced by iota-toxin. Furthermore, treatment of the cells with Ib resulted in the production of ceramide in cytoplasmic vesicles. These observations showed that ASMase promotes the internalization of iota-toxin into target cells.
Sigma-Aldrich
Bromoenol lactone, ≥98% (TLC)