Merck
  • Identification of a new member of the tumor necrosis factor family and its receptor, a human ortholog of mouse GITR.
Identification of a new member of the tumor necrosis factor family and its receptor, a human ortholog of mouse GITR.
Current biology : CB (1999-03-13)
A L Gurney, S A Marsters, R M Huang, R M Pitti, D T Mark, D T Baldwin, A M Gray, A D Dowd, A D Brush, A D Heldens, A D Schow, A D Goddard, W I Wood, K P Baker, P J Godowski, A Ashkenazi
ABSTRACT
The tumor necrosis factor (TNF) and TNF receptor (TNFR) gene superfamilies regulate diverse biological functions, including cell proliferation, differentiation, and survival [1] [2] [3]. We have identified a new TNF-related ligand, designated human GITR ligand (hGITRL), and its human receptor (hGITR), an ortholog of the recently discovered murine glucocorticoid-induced TNFR-related (mGITR) protein [4]. The hGITRL gene mapped to chromosome 1q23, near the gene for the TNF homolog Fas/CD95 ligand [5]. The hGITR gene mapped to chromosome 1p36, near a cluster of five genes encoding TNFR homologs [1] [6]. We found hGITRL mRNA in several peripheral tissues, and detected hGITRL protein on cultured vascular endothelial cells. The levels of hGITR mRNA in tissues were generally low; in peripheral blood T cells, however, antigen-receptor stimulation led to a substantial induction of hGITR transcripts. Cotransfection of hGITRL and hGITR in embryonic kidney 293 cells activated the anti-apoptotic transcription factor NF-kappaB, via a pathway that appeared to involve TNFR-associated factor 2 (TRAF2) [7] and NF-kappaB-inducing kinase (NIK) [8]. Cotransfection of hGITRL and hGITR in Jurkat T leukemia cells inhibited antigen-receptor-induced cell death. Thus, hGITRL and hGITR may modulate T lymphocyte survival in peripheral tissues.
Sigma-Aldrich
AITRL human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture