Merck
  • Analysis of 10 β-agonists in pork meat using automated dispersive pipette extraction and LC-MS/MS.
Analysis of 10 β-agonists in pork meat using automated dispersive pipette extraction and LC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2018-03-24)
Kaylee R Mastrianni, Kalmorat Metavarayuth, William E Brewer, Qian Wang
ABSTRACT
An analytical procedure for the analysis of 10 β-adrenergic agonists (cimaterol, terbutaline, salbutamol, isoxsuprine, ractopamine, cimbuterol, clenbuterol, brombuterol, mabuterol and mapenterol) in pork meat was developed and validated using LC-MS/MS. An automated dispersive pipette extraction (DPX) was employed on a Hamilton Microlab® NIMBUS96® platform to extract the analytes of interest prior to LC-MS/MS analysis. The extraction time was <20 min with a total LC-MS/MS run time of 9.6 min. The method was fully validated in accordance with the international guidelines (European Commission Decision 2002/657/EC and National Standards of People's Republic of China, GB/T 22286-2008) for limit of detection, limit of quantitation, carryover, extraction efficiency, matrix effects, linearity, and within and between-run precision. The proposed method can be successfully used in the routine determination of 10 β-adrenergic agonists in pork and as a potential solution for compliance monitoring in regulatory laboratories.
Product Description
Supelco
Cimaterol, VETRANAL®, analytical standard
Supelco
Cimbuterol, VETRANAL®, analytical standard
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)