Merck
  • 14-3-3ζ promotes esophageal squamous cell carcinoma invasion by repressing S1PR2 protein expression through NF-κB signaling.
14-3-3ζ promotes esophageal squamous cell carcinoma invasion by repressing S1PR2 protein expression through NF-κB signaling.
Archives of biochemistry and biophysics (2018-02-20)
Song Tong, Sheng-Cai Chen, Kai-Ying Xu, Bin Fang, Si-Hua Wang, Jian-Jun Wang
ABSTRACT
14-3-3ζ is overexpressed in several cancers, including esophageal squamous cell carcinoma (ESCC), and plays an important role in tumorigenesis. However, the mechanisms underlying its tumorigenesis remain unclear. Here we report that 14-3-3ζ was upregulated in ESCC tumors, compared with adjacent normal tissues; 14-3-3ζ levels were positively correlated with ESCC lymph node metastasis and recurrence. Overexpression of 14-3-3ζ promoted the tumor growth and invasion of ESCC in vitro and in vivo, whereas depletion of 14-3-3ζ suppressed these effects. Moreover, 14-3-3ζ reduces expression of genes mediating S1P/S1PR2 signaling, and this effect is mediated through activation of NF- κ B. Taken together, 14-3-3ζ contributes to ESCC tumorigenesis and progression through repressing S1PR2 signaling and may act as a new therapeutic target for ESCC.
Sigma-Aldrich
Caffeic acid phenethyl ester, ≥97% (HPLC), powder
Sigma-Aldrich
JSH-23, ≥98% (HPLC), solid