![Zoledronic Acid Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/513/243/10de8edb-d031-491e-b4d8-2a98dd926cc5/640/10de8edb-d031-491e-b4d8-2a98dd926cc5.jpg)
Zoledronic acid Related Compound A
![Zoledronic Acid Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/513/243/10de8edb-d031-491e-b4d8-2a98dd926cc5/640/10de8edb-d031-491e-b4d8-2a98dd926cc5.jpg)
Synonym(s):
2-(1H-Imidazole-1-yl)acetic acid
Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2