![X-Gal OmniPur® Grade](/deepweb/assets/sigmaaldrich/product/structures/247/263/57654e94-ae59-485c-ae90-4a5da7fb51a9/640/57654e94-ae59-485c-ae90-4a5da7fb51a9.png)
X-Gal
![X-Gal OmniPur® Grade](/deepweb/assets/sigmaaldrich/product/structures/247/263/57654e94-ae59-485c-ae90-4a5da7fb51a9/640/57654e94-ae59-485c-ae90-4a5da7fb51a9.png)
Synonym(s):
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, 5-Bromo-4-chloro-3-indolyl β-D-galactoside, BCIG, X-Gal, OmniPur® X-Gal
Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing