Skip to Content
Merck
Whiskey lactone ≥98%, FG

Whiskey lactone

Whiskey lactone ≥98%, FG
Synonym(s):
(±)-5-Butyl-4-methyldihydro-2(3H)-furanone
Empirical Formula (Hill Notation):
C9H16O2
CAS Number:
Molecular Weight:
156.22
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing