![Vecuronium bromide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/297/547/fdd14bdc-6006-419f-8ddd-392a45a6922d/640/fdd14bdc-6006-419f-8ddd-392a45a6922d.png)
Vecuronium bromide
![Vecuronium bromide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/297/547/fdd14bdc-6006-419f-8ddd-392a45a6922d/640/fdd14bdc-6006-419f-8ddd-392a45a6922d.png)
Synonym(s):
1-[(2β,3α,5α,16β,17β)-3,17-Bis(acetyloxy)-2-(1-piperidinyl)androstan-16-yl]-1-methyl-piperidinium bromide, Vecuronium bromide
Empirical Formula (Hill Notation):
C34H57BrN2O4
CAS Number:
Molecular Weight:
637.73
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing