Urea Related Compound A
Synonym(s):
Biuret, Allophanic acid amide, Carbamoylurea
Linear Formula:
NH2CONHCONH2
CAS Number:
Molecular Weight:
103.08
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing