Skip to Content
Merck
Tris(4-tert-butylphenyl)sulfonium perfluoro-1-butanesulfonate electronic grade, ≥99% trace metals basis

Tris(4-tert-butylphenyl)sulfonium perfluoro-1-butanesulfonate

Tris(4-tert-butylphenyl)sulfonium perfluoro-1-butanesulfonate electronic grade, ≥99% trace metals basis
Linear Formula:
[(CH3)3C6H4]3SC4F9SO3
CAS Number:
Molecular Weight:
730.79

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group