Skip to Content
Merck
Triasulfuron PESTANAL®, analytical standard

Triasulfuron

Triasulfuron PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C14H16ClN5O5S
CAS Number:
Molecular Weight:
401.83

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing