Skip to Content
Merck
Thiazole ≥99%

Thiazole

Thiazole ≥99%
Empirical Formula (Hill Notation):
C3H3NS
CAS Number:
Molecular Weight:
85.13
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing