![Thiamazole impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/295/188/3d91cd0b-e777-4efb-88e3-991341848eb4/640/3d91cd0b-e777-4efb-88e3-991341848eb4.png)
Thiamazole impurity A
![Thiamazole impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/295/188/3d91cd0b-e777-4efb-88e3-991341848eb4/640/3d91cd0b-e777-4efb-88e3-991341848eb4.png)
Synonym(s):
(Methylamino)acetaldehyde dimethyl acetal, 1,1-Dimethoxy-2-methylaminoethane, 2,2-Dimethoxy-N-methylethylamine
Linear Formula:
CH3NHCH2CH(OCH3)2
CAS Number:
Molecular Weight:
119.16
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing