Tenofovir disoproxil Related Compound B
Synonym(s):
(E)-9-(Prop-1-enyl)-9H-purin-6-amine
Empirical Formula (Hill Notation):
C8H9N5
CAS Number:
Molecular Weight:
175.19
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2