Skip to Content
Merck
Tegafur ≥98% (HPLC), powder

Tegafur

Tegafur ≥98% (HPLC), powder
Synonym(s):
5-Fluoro-1-(tetrahydro-2-furyl)uracil, Ftorafur, Tegafur
Empirical Formula (Hill Notation):
C8H9FN2O3
CAS Number:
Molecular Weight:
200.17
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing