![Tamsulosin Racemate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/913/763/10fb7441-9b31-4a63-a6dc-0a24bcd71662/640/10fb7441-9b31-4a63-a6dc-0a24bcd71662.png)
Tamsulosin Racemate
![Tamsulosin Racemate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/913/763/10fb7441-9b31-4a63-a6dc-0a24bcd71662/640/10fb7441-9b31-4a63-a6dc-0a24bcd71662.png)
Synonym(s):
5-[2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide hydrochloride
Empirical Formula (Hill Notation):
C20H28N2O5S · HCl
CAS Number:
Molecular Weight:
444.97
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing