Sodium phosphate dibasic solution
Synonym(s):
Disodium hydrogen phosphate solution, di-Sodium hydrogen phosphate solution
Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing