Skip to Content
Merck
Potassium palmitate-1,2,3,4-13C4 endotoxin tested, 99 atom % 13C

Potassium palmitate-1,2,3,4-13C4

Potassium palmitate-1,2,3,4-13C4 endotoxin tested, 99 atom % 13C
Synonym(s):
Hexadecanoic acid-1,2,3,4-13C4 potassium salt, Palmitic acid-1,2,3,4-13C4 potassium salt, Potassium hexadecanoate-1,2,3,4-13C4
Linear Formula:
CH3(CH2)11(13CH2)313CO2K
CAS Number:
Molecular Weight:
298.49

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Special Grade