Skip to Content
Merck
Potassium oleate-13C18 endotoxin tested, 99 atom % 13C, 98% (CP)

Potassium oleate-13C18

Potassium oleate-13C18 endotoxin tested, 99 atom % 13C, 98% (CP)
Synonym(s):
Oleic acid-13C18 potassium salt
Linear Formula:
13CH3(13CH2)713CH=13CH(13CH2)713CO2K
Molecular Weight:
338.42

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Isotope
Markush Class
Markush Group
Showing 1-2 of 2