Skip to Content
Merck
Phosphoenol-pyruvate 97% (PEP-K), pkg of 1 g

Phosphoenol-pyruvate

Phosphoenol-pyruvate 97% (PEP-K), pkg of 1 g
Synonym(s):
Phospho(enol)pyruvic acid monopotassium salt, 2-(Phosphonooxy)-2-propenoic acid monopotassium salt, mono-Potassium phosphoenolpyruvate, PEP-K
Empirical Formula (Hill Notation):
C3H4KO6P
CAS Number:
Molecular Weight:
206.13

Products

Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing