![Phenylalanine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/383/821/2714fb32-3e3b-4b81-83d2-248fbb6f95f8/640/2714fb32-3e3b-4b81-83d2-248fbb6f95f8.png)
Phenylalanine
![Phenylalanine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/383/821/2714fb32-3e3b-4b81-83d2-248fbb6f95f8/640/2714fb32-3e3b-4b81-83d2-248fbb6f95f8.png)
Synonym(s):
L-Phenylalanine, (S)-2-Amino-3-phenylpropionic acid
Linear Formula:
C6H5CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
165.19
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing