Parteck® SI 450 (Sorbitol)
Synonym(s):
Parteck® Sorbitol
Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing