Skip to Content
Merck
Padimate O United States Pharmacopeia (USP) Reference Standard

Padimate O

Padimate O United States Pharmacopeia (USP) Reference Standard
Synonym(s):
2-Ethylhexyl 4-(dimethylamino)benzoate
Linear Formula:
(CH3)2NC6H4CO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
277.40

Products

Boiling Point (°C)
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique