Skip to Content
Merck
Oryzalin PESTANAL®, analytical standard

Oryzalin

Oryzalin PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C12H18N4O6S
CAS Number:
Molecular Weight:
346.36
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing