![Orphenadrine for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/573/250/f5e8b963-d13f-46fa-b493-63ca6b58db1f/640/f5e8b963-d13f-46fa-b493-63ca6b58db1f.png)
Orphenadrine for peak identification
![Orphenadrine for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/573/250/f5e8b963-d13f-46fa-b493-63ca6b58db1f/640/f5e8b963-d13f-46fa-b493-63ca6b58db1f.png)
Synonym(s):
Orphenadrine hydrochloride, β-Dimethylaminoethyl 2-methylbenzhydryl ether hydrochloride, N,N-Dimethyl-2-(2-methylbenzhydryloxy)ethylamine hydrochloride, o-Methyldiphenhydramine hydrochloride
Empirical Formula (Hill Notation):
C18H23NO · HCl
CAS Number:
Molecular Weight:
305.84
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing